Would you like to log in again?
Oxoid VCAT Selective Supplement, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SR104B SR104B
89423-162EA 604.72 USD InStock
CA89423-162 89423-162
Oxoid VCAT Selective Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic selective supplement for culture media, for the isolation of Neisseria gonorrhoeae
An antibiotic selective supplement for culture media, for the isolation of Neisseria gonorrhoeae, such as Thayer Martin. Contains Vancomycin, Colistin, Anisomysin, and Trimethoprim.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty