Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Corning® Collection Containers, Corning
Supplier: Corning
Corning®
91-200-20 91-200-10 91-200-01 91-200-02 91-200-48 91-100-30 91-200-36 91-200-10 91-200-45 91-200-48 91-200-05 91-200-82 91-200-43 91-200-01 91-200-42 91-200-47 91-200-41 91-200-02 91-200-42 91-200-45 91-200-39 91-200-20 91-200-39 91-200-36 91-200-82 91-200-83 91-200-41 91-200-83 91-002-MX 91-200-47 91-200-43 91-200-05
89423-196EA 71.58 USD InStock
89423-196 89423-198 89423-192 CA89423-204 89423-236 89423-214 CA89423-202 89423-216 89423-238 89423-210 CA89423-200 CA89423-220 89423-212 CA89423-218 89423-208 CA89423-216 CA89423-238 89423-190 CA89423-196 89423-220 89423-204 CA89423-236 CA89423-214 CA89423-212 89423-206 89423-200 CA89423-210 89423-202 CA89423-198 CA89423-208 89423-218 CA89423-206
Corning® Collection Containers, Corning
Bags Media Bags
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty