Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Corning® Tank Liners, Corning
Supplier: Corning
Corning®
91-300-20 91-300-15 91-300-60 91-300-80 91-300-60 91-300-80 91-300-30 91-300-20 91-300-30 91-300-35 91-300-25 91-300-35 91-300-15 91-300-25
89423-262EA 134.16 USD InStock
89423-262 CA89423-262 89423-264 CA89423-260 10770-488 89423-260 CA10770-488 CA89423-258 89423-258 CA89423-256 89423-266 CA89423-266 CA89423-264 89423-256
Corning® Tank Liners, Corning
Bags Media Bags
Corning® tank liners are designed to fit cylindrical tanks.
  • Available in a range of sizes
Tank liners are manufactured from LDPE.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty