Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner bio-one
Vacuette®
459001 459000
89423-412CS 490.09 USD InStock
89423-412 75796-090
VACUETTE® Secondary Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
VACUETTE® Secondary Tubes are used as a secondary container for aliquoting, storing, and transporting blood, blood components, and urine from the primary tube in the clinical laboratory.
  • Single-use tubes
  • Nonsterile
  • Used for analysis on laboratory analyzers
They may also be used for analysis on laboratory analysers. The choice of tube depends on the analyser and the intended use. These single-use tubes are nonsterile and are not evacuated.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty