Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Compact Cart System, Micronova

Supplier: Micronova
Slimline™ SlimT™
B-99 W-9 W-9B C-92 RP-05 C-91 C-93 W-92
68400-672EA 430.91 USD InStock
68400-672 89413-040 89140-710 89423-424 94000-434 89423-420 89423-430 89423-422
Stainless Steel Compact Cart System, Micronova
Buckets
Compact and highly manoeuvrable stainless steel cart system with smooth, stainless steel buckets and 360° rotation Prevenz Antimicrobial Wheels. Available in single, double and triple bucket configurations.

  • Durable bacteria-resistant construction
  • Prevenz Antimicrobial Wheels with 360° rotation
  • Smooth bucket interiors with no ridges or seams to harbor bacteria
  • Compact size allows for easy storage
  • Generous width easily accommodates 45.7 cm (18") mops
  • Foot activated brake for stability while cleaning
  • Bucket includes a stainless steel carrying handle
  • Wringers attach directly to rim of bucket
  • Optional wringer bar allows W-6 wringer to be positioned above the rinse bucket
  • Entire unit is autoclavable

The Micronova Stainless Steel Compact Cart System features a bacteria-resistant construction with antimicrobial wheels, smooth, molded stainless steel buckets with no ridges or seams and a compact, stainless steel frame. Compatible with MegaWringer™ and Slim-T™ series stainless steel mop wringers. The C3A-03 Wringer Bar attachment positions the W-6 Downpress Wringer above the rinse bucket or empty third bucket for simplified wringing.

Ordering information: Buckets are included with carts. All other components are optional and must be ordered separately.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty