Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sliding Drawer Freezer Racks for Tubes, Thermo Scientific
Supplier: Thermo Scientific
1950818 1950786 1950792 1950802 1950799 1950796 1950819 1950816 1950800 1950780 1950782 1950787 1950798 1950784 1950791
89423-556EA 386.76 USD InStock
89423-556 89423-512 89423-522 89423-514 89423-524 89423-530 89423-540 89423-532 89423-510 89423-564 89423-516 89423-526 89423-548 89423-518 89423-528
Sliding Drawer Freezer Racks for Tubes, Thermo Scientific
Racks Cooling Racks
Maximize tube storage capacity per freezer with Thermo Scientific™ sliding drawer racks for tubes (four and five inner door freezers), a racking solution for five inner-door, ultra-low temperature freezers.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Racking solutions are designed to hold tubes ranging from 200 µL up to 5 mL, offering maximized sample storage capacity for both five and four inner door freezers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty