Would you like to log in again?
Uvex Carbonvision™ Safety Goggles, Honeywell Safety
Supplier: Honeywell Safety
uvex carbonvision™
S1650D S1650DF S1651D S1651DF
89423-592CS 2133.35 USD InStock
89423-592 89423-594 89423-596 89423-598
Uvex Carbonvision™ Safety Goggles, Honeywell Safety
Goggles
These safety goggles with Dura-streme® dual anti-fog/anti-scratch coating are designed with flexible soft frame which adapts perfectly to the face to ensure a perfect fit and seal around the eyes for high-performance protection and all-day comfort.
  • Soft, flexible body which is permanently fixed to the frame, making it easy to clean and maintain
  • Protects from impact, dust/airborne particles as well as chemical splash hazards
  • Convenient clip makes it easy to remove headband before cleaning
Low profile and its compact design is ideal for wearing with disposable respirators and protective caps. High-quality headband is available in neoprene or fabric and offers a comfortable fit and long-lasting durability. Available in Clear and Gray lens tints.
Certifications: Meets the ANSI Z87.1-2010 standard. Certified to the requirements of the CSA Z94.3 standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty