Would you like to log in again?
Tuerk solution
Supplier: AZER SCIENTIFIC INC
Synonyms: Acetic acid gentian violet solutionTurk solution
ES930 ES932 ES931
89423-628EA 34.78 USD InStock
89423-628 89423-632 89423-630
Tuerk solution
Tuerk solution
Turk's blood diluting fluid has a stain (gentian violet) plus 6% acetic acid and is ideal for manual leukocyte counting.
  • Ideal for Manual Leukocyte Counting
  • Six Percent Acetic Acid
  • Gentian Violet Stain
Stains the nuclei of the white blood cells making them easier to see and count.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty