Would you like to log in again?
BCSA, Selective for Burkholderia cepacia
Supplier: Hardy Diagnostics
G09 GA09 G09
89423-720PK 84.87 USD InStock
CA89423-720 89423-720 76288-334
BCSA, Selective for Burkholderia cepacia
Media for Microbiology Dehydrated Media for Microbiology
Burkholderia cepacia Selective Agar is ideal for the selective isolation and differentiation of Burkholderia (Pseudomonas) cepacia from sputum samples from cystic fibrosis patients and other clinical specimens.
Packaging: Pack of 10.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty