Would you like to log in again?
Capto™ adhere ImpRes Multimodal Chromatography Media, Cytiva
Supplier: Cytiva
Capto™
17371502 17371501 17-3715-01 17-3715-02
89425-702EA 321.07 USD InStock
89425-702 89425-704 CA11027-082 CA11027-084
Capto™ adhere ImpRes Multimodal Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Capto™ adhere ImpRes is a strong anion exchange multimodal BioProcess chromatography medium (resin), designed to meet the demands of modern large-scale manufacturers, for high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules. Capto™ adhere ImpRes is designed to allow effective MAb polishing in the second or third step of a purification scheme downstream of the protein A capture step. Polishing can be performed in either bind and elute (binding) or flowthrough (non binding) modes.
  • High yields achieved through the high-resolution beads and selectivity of the ligand
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of a platform approach to MAb process development
  • Allows separation of MAb charged variants
The high selectivity of the anion multimodal ligand allows operation in a variety of process conditions to solve challenging purification problems. Contaminants such as DNA, host cell proteins, leached protein A, aggregates, and viruses are efficiently removed. The multimodal anion exchanger ligand of Capto adhere ImpRes is immobilized to the base matrix and interacts with the target molecule through multiple types of interactions. Capto adhere ImpRes is available in a range of different bulk pack sizes for easy scale-up and pre-packed formats for process development. As member of the BioProcess media range, Capto adhere ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty