Would you like to log in again?
SCREEN-WELL® Autophagy Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2837-0500 BML-2837-0100
89426-196EA 3874.71 USD InStock
89426-196 89426-194
SCREEN-WELL® Autophagy Library, Enzo Life Sciences
Cell Culture Supplements and Additives
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and in vitro applications.
The Screen-well Autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications.
Ordering information: Contains 94 compounds with defined autophagy-inducing or -inhibitory activity.  A variety of structurally and mechanistically different compound classes are included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty