Would you like to log in again?
Anaerobe Jar, Hardy Diagnostics
Supplier: Hardy Diagnostics
16000 16000
89426-216EA 518.41 USD InStock
CA89426-216 89426-216
Anaerobe Jar, Hardy Diagnostics
Jars General Use Jars
This anaerobic jar is specially formulated for temperature stability and durability
It’s unbreakable, lightweight, transparent and, of course, autoclavable. Double screw clamp, coupled with the patented Quad Ring, ensures a more positive seal, superior to conventional O-rings. It also doubles as a handy carrying handle. The 3/8” thick anodized aluminum lid won’t warp, and it’s guaranteed to resist cracking or breaking for ten years.
Delivery information: Shipped complete with a stainless steel rack to accommodate 100 mm petri dishes and a stainless steel catalyst basket.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty