Would you like to log in again?
McFarland Standard
Supplier: Hardy Diagnostics
ML3 ML3 ML6 ML5 ML5 ML8 ML1 ML2 ML4 ML7 ML8 ML05 ML4 ML7 ML05 ML2 ML1 ML6
89426-224EA 77.49 USD InStock
CA89426-224 89426-224 89426-234 CA89426-226 89426-222 CA89426-228 89426-232 89426-220 CA89426-218 89426-230 CA89426-230 89426-218 CA89426-220 89426-228 CA89426-232 CA89426-222 89426-226 CA89426-234
McFarland Standard
Media for Microbiology Dehydrated Media for Microbiology
The McFarland Latex Standards are prepared from suspensions of uniform latex particles with similar absorbance values as the original barium sulfate standards.
The stability of McFarland Latex Standards allows for a significantly longer shelf-life than the original barium sulfate standards due to reduced light sensitivity.
Latex Equivalent ML05, #1, #2, #3, #4, #5, #6, #7, and #8 sold separately.
Latex Equivalent ML05, #1, #2, #3, #4, #5, #6, #7, and #8 sold separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty