Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bile HardyDisks™, Hardy Diagnostics
Supplier: Hardy Diagnostics
HardyDisks™
Z7091 Z7091
89426-240EA 84.99 USD InStock
89426-240 CA89426-240
Bile HardyDisks™, Hardy Diagnostics
Microbiology Test Systems Endotoxin Tests
Bile HardyDisks™, to aid in the identification of Bacteroides fragilis group on the basis of bile resistance, 1 x 50 disks by Hardy Diagnostics, freeze on arrival.
Differentiates Bacteroides fragilis group from other anaerobic bacteria. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Differentiates Bacteroides fragilis group from other anaerobic bacteria
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty