Would you like to log in again?
Bushing Adapter, Glassco
Supplier: Glassco
013.478.02 013.478.01
89426-322EA 30.34 USD InStock
89426-322 89426-320
Bushing Adapter, Glassco
Adapters Adapters, Conical Ground Joints
Bushing adapters allow for compact assemblies of dissimilar joints.
  • Top [ST] joint
  • Bottom [ST] joint
  • Manufactured from borosilicate 3.3 glass
Bushing-style adapters have upper and lower standard taper joints.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty