Would you like to log in again?
Vacuum Adapter, with Outer Joint, Glassco
Supplier: Glassco
020.470.03
89426-348EA 29.5 USD InStock
89426-348
Vacuum Adapter, with Outer Joint, Glassco
Adapters Adapters, Conical Ground Joints
This vacuum adapter features an outer standard taper joint.
  • Top outer [ST] joint
  • Manufactured from borosilicate 3.3 glass
Adapter is ideal for use as a vent, or for the introduction of pressure or vacuum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty