Would you like to log in again?
West Condenser, Glassco
Supplier: Glassco
182.470.02B 182.470.01 182.470.00 182.470.03 182.470.06 182.470.02A 182.470.02
89426-654EA 86.83 USD InStock
89426-654 89426-664 89426-660 89426-670 89426-658 89426-668 89426-666
West Condenser, Glassco
Condensers
Condensers feature narrow annular space for fast coolant speed, thus ensuring high cooling efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty