Would you like to log in again?
Condenser for Soxhlet Extractors, Glassco
Supplier: Glassco
191.470.02
89426-690EA 160.93 USD InStock
89426-690
Condenser for Soxhlet Extractors, Glassco
Condensers
Allihn reflux condenser with a water cooled inner drip joint at bottom
Made from Borosilicate 3.3 glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty