Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
RO Series Multi-Position Stirrer Plates, IKA Works
Supplier: IKA
0003691000 0003690500 0003692500
89426-972EA 4929.51 USD InStock
89426-972 89426-970 89426-968
RO Series Multi-Position Stirrer Plates, IKA Works
Stirrers Magnetic Hotplate Stirrers, Multi-position
5, 10, or 15-position digital magnetic stirrer without heating, designed for synchronous stirring
The magnetic coil technology provides noiseless and consistent stirring on all positions. Speed remains constant, even when load changes. The closed and compact design allows easy cleaning and protects the equipment against the penetration of liquids. Additionally, a removable transparent anti-slip mat is included.
Certifications: UL, cUL, CSA, CE certified, RoHS compliant.
Ordering information: Unit comes with detachable power cord. Three year mnaufacturer's warranty with product registration.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty