Would you like to log in again?
Incubator Shaker, 230 V, KS 3000i control
Supplier: IKA
3940000
89426-976EA 9815.75 USD InStock
89426-976
Incubator Shaker, 230 V, KS 3000i control
Shakers and Mixers Shaking Incubators
Innovatively designed, compact incubator shaker allows safe, unattended operation in a temperature-controlled environment.
  • 230 V model
The Incubator Shaker KS 3000i Control features controls with antimicrobial coating for reduction of bacteria, and a junction box in the workspace for connection of an additional temperature sensor, e.g. PT-1000 (included with delivery). The wide range of attachments allows for using almost all shapes and sizes of vessels. Unit stops automatically when hood is lifted. A collecting tray with drain hose is on rear of the unit. Electronic timer also features error code display.
The unit has a large LED display for speed, temperature, and timer. An integrated PID temperature control allows the use of PT 1000 temperature sensors to assure highly accurate temperature control. The electronically controlled incubator comes equipped with an RS232 interface and USB ports for use with a PC. All functions can be controlled and documented using labworldsoft® (software sold separately).
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with three-year manufacturer's warranty. Attachments and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89426-974) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty