Would you like to log in again?
Incubator Shaker, 230 V, KS 3000ic control
Supplier: IKA
3940100
89426-980EA 12274.19 USD InStock
89426-980
Incubator Shaker, 230 V, KS 3000ic control
Shakers and Mixers Shaking Incubators
Compact incubator shaker features a built-in cooling coil for connection to an external cooling unit.
  • 230 V model
The Incubator Shaker KS 3000ic Control featurers controls with antimicrobial coating for reduction of bacteria, and a junction box in the workspace for connection of an additional temperature sensor, e.g. PT-1000 (included with delivery). The wide range of attachments allows for using almost all shapes and sizes of vessels. Unit stops automatically when hood is lifted. A collecting tray with drain hose is on rear of the unit. Electronic timer also features an error code display.
Designed for safe, unattended operation in a temperature-controlled environment. The unit has a large LED display for speed, temperature, and timer. Integrated PID temperature control allows the use of PT 1000 temperature sensors to assure highly accurate temperature control. The electronically controlled incubator comes equipped with an RS 232 interface and USB ports for use with a PC. All functions can be controlled and documented using labworldsoft® (software sold separately).
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with three-year manufacturer's warranty. Attachments and other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89426-978) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty