Would you like to log in again?
Spor-Klenz® Ready To Use ETO Processed Packaged Cold Sterilant, STERIS®
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
652873
89426-982CS 527.76 USD InStock
89426-982
Spor-Klenz® Ready To Use ETO Processed Packaged Cold Sterilant, STERIS®
Cleaning Agents Disinfectants
Spor-Klenz® Ready-To-Use ETO process packaged sterilants for sterilization and disinfection of environmental, hard non-porous surfaces.
  • Ethylene oxide (ETO) processed through a validated process for easy introduction into your aseptic areas
  • Fast acting, specifically formulated with liquid sporicides
  • Excellent material compatibility with low toxicity profile
It is blend of peracetic acid, hydrogen peroxide and acetic acid that provides fast, broad spectrum efficacy against bacteria,
fungi, viruses and bacterial spores. This formulation offers operational flexibility by being diluted with water for use as a disinfectant or sanitizer. These products have an extensive documentation package to meet your validation objectives.
Certifications: Manufactured in accordance with cGMP and USP sterility tested.
Packaging: 0.2 micron filtered, double bagged and packed 6 bottles per case.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty