Would you like to log in again?
Phalloidin Conjugates, Biotium
Supplier: Biotium
00042-T 00052-T 00043-T 00053 00044-T 00055-T 00047 00053-T 00050-T 00045-T 00054 00046 00050 00055 00043 00064 00030 00041 00040 00051-T 00040-T 00033 00054-T 51 00064-T 00048 00044 00052 00032 00049 00042 00034 00045 00028 00041-T 00046-T 00027 00048-T 00049-T 00047-T 00034-T
76403-876EA 527.57 USD InStock
76403-876 89138-128 89138-126 89138-146 89138-124 76403-878 89138-122 89138-144 89138-120 89138-142 89138-140 89427-148 89427-126 89427-128 89427-134 89427-136 75784-078 89427-152 76458-726 89427-130 89427-132 71004-008 76458-724 89427-150 75784-076 89138-118 75784-074 89138-138 89138-116 89138-136 89138-134 89138-132 89138-130 89427-138 89427-144 89427-146 89427-086 89427-140 89427-142 10018-144 75784-080
Phalloidin Conjugates, Biotium
Protein Linking and Labeling Systems
A highly selective actin filament stain for fixed and permeabilized cells. Labeled phalloidins have similar affinity for both large and small filaments.
  • Choice of 18 bright and photostable CF® dyes, biotin, or other common fluorophores
  • Dye options for super-resolution and 2-photon imaging
  • Excellent signal to background with negligible non-specific staining
Biotium offers a variety of phalloidin products, including conjugates of our outstanding CF™ dyes, which offer advantages in brightness, photostability, and/or signal-to-noise ratio compared to other fluorescent dyes. Please see the CF™ dye reference chart under the Datasheets tab to select the appropriate CF™ dye for your application.
It is a bicyclic peptide that binds specifically to F-actin. Fluorescent conjugates of phalloidin are convenient tools to investigate the distribution of F-actin in fixed and permeabilized cells or tissue sections. One unit (U) of phallodin is sufficient to stain one sample (200µL staining volume).
Fluorescently-labeled phalloidins bind to F-actin with nanomolar affinity. Labeled phalloidins have similar affinity for both large and small filaments, binding in a stoichiometric ratio of about one phalloidin molecule per actin subunit in muscle and nonmuscle cells from various species of plants, animals and fungi. Different from antibodies, the binding affinity of phalloidin does not change significantly with actin among different species.
Fluorescently-labeled phalloidin is typically used to stain fixed and permeabilized cells. Labeled phalloidins are not cell-permeant and have therefore has not been used extensively with living cells, however they can be loaded into live cells via cationic liposomes.
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Ordering information: Supplied as lyophilized solid in 50 U and 300 U sizes. Typical staining concentration for F-actin in fixed and permeabilized cells is 5 U/ml.
Packaging: Phalloidin stock solutions are very stable. But binding of fluorescent phalloidins to cells is more labile than other probes, such as antibodies, and staining can become non-specific or lost over time. For best results, store phalloidin-stained samples in a suitable mounting medium at 4 °C, protected from light. For certain phalloidin conjugates, especially CF®405M, CF®647, and CF®680, Biotium recommends imaging immediately or shortly after staining. Staining with Biotium's other CF® dye phalloidins usually is stable for up to a week when specimens are stored at 4 °C, protected from light.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty