Would you like to log in again?
Adjustable Right Angle Clamp Holder
Supplier: HUMBOLDT MFG CO
H-8342 H-8342
470005-376EA 28.05 USD InStock
470005-376 89427-302
Adjustable Right Angle Clamp Holder
Clamps General Purpose Clamps
Sturdy cast aluminum alloy clamp can be swiveled and locked at any desired angle.
The clamp can be attached to rods up to 13 mm ( 1/2") in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty