Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Puritan® Dry Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
25-8062PDBT 25-8061PCBT 25-8061PABT 25-8061PRBT 25-8061PDBT 25-8061WCBT 25-8062PRBT 25-15062PFBT 25-15061PFBT
89427-672CS 426.47 USD InStock
89427-672 75791-992 89427-674 75791-993 10119-860 89427-670 75788-572 89427-668 10119-856
Puritan® Dry Transport Systems, Puritan Medical Products
Swabs Transport Swabs
Get all your specimen collection devices in one place
Safe, sterile specimen transport. No matter which swab(s) are chosen, all of the transport systems include a round-bottom tube made from virgin polypropylene resin, which will securely hold the swab. The system maintains a safe and sterile environment.
Tubes include a distinctive label detailing product identity, expiry date, patient identification, and collection information.
Dry Transport Systems are ideal for the collection and transport of specimen when no medium is required. Choose from a range of swab types with rayon, polyester, foam, HydraFlock®, or PurFlock Ultra® tips. Single or double options available.
Certifications: Made in the USA. CE marked.
Ordering information: Double configurations are individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty