Would you like to log in again?
Microwave Reaction Vials, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5704-10 5704-05 5704-02
89427-730CS 882.47 USD InStock
89427-730 89427-728 89427-726
Microwave Reaction Vials, Ace Glass Incorporated
Vials
Heavy wall, Type 1 clear borosilicate glass for higher pressure, microwave digestion.
20 mm flange top to accomodate crimp seals. Available with either conical or rounded bottoms.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty