Would you like to log in again?
Baffle for Temperature Probe, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
12193-07 12193-09 12193-03
89427-774EA 947.64 USD InStock
89427-774 89427-776 89427-778
Baffle for Temperature Probe, PTFE, Ace Glass Incorporated
Stirrers Stirring Tools
A PTFE baffle with an integral stainless steel tube
Can be used for either liquid addition into a reactor or primarily a temperature probe. The baffle will accomodate a 1/4" OD thermocouple. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or thermocouple. Thermocouple not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty