Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum Seals, 20 mm, Lined, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5708-38 5708-06 5708-18 5708-33 5708-05 5708-34 5708-31 5708-20 5708-37 5708-11 5708-10 5708-35 5708-32 5708-19 5708-12 5708-36 5708-30
89427-806PK 202.1 USD InStock
89427-806 89427-808 89097-130 89427-802 89427-804 89427-798 89427-800 89427-794 89097-138 89097-128 89427-796 89097-136 89097-126 89097-134 89097-124 89097-132 89427-792
Aluminum Seals, 20 mm, Lined, Ace Glass Incorporated
Closures Chromatography Vial Closures
These seals are 20mm lined, natural aluminum crimp style seals for headspace, serum and other styles of vials with 20mm crimp finish tops

Available in natural aluminum with various PTFE/rubber linings. Colored aluminum available via special order. A magnetic crimp seal version is available for use with robotics and a pressure release version designed to release at 3 Bar (43 psi) pressure.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty