Would you like to log in again?
Raschig Rings, Ace Glass
Supplier: Ace Glass
8033-08 8033-06 8033-04 8033-10
89427-870EA 333.13 USD InStock
89427-870 89427-872 89427-868 89427-866
Raschig Rings, Ace Glass
Distillation Column Packing
Raschig rings fabricated from borosilicate glass.
Wall weight on 5 mm size is approximately 0.8 mm, other sizes have wall weight of approximately 1.0 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty