Would you like to log in again?
Filtration Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3709-05 3708-10 3708-02 3708-20 3708-16 3709-18 3709-16 3708-14
89427-882EA 2025.56 USD InStock
89427-882 89428-174 89427-884 89428-178 89428-176 89427-890 89427-886 89427-888
Filtration Apparatus, Ace Glass Incorporated
Filters Membrane Filter Units
This borosilicate glass filtration apparatus accomodates 75 mm membrane filters.
The new design utilizes a PTFE Ace-Thred adapter to join the glass funnel, filter flask and fritted disc together. This design eliminates the need for the clamping device, stopper and base. The unit is easy to assemble, disassemble and clean. The fritted disc can be easily replaced and other porosity discs used. The apparatus is easier to use and more economical than old designs.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty