Would you like to log in again?
PTFE [ST] Joint Inserts for Flat Glass Heads, Ace Glass Incorporated
Supplier: Ace Glass
6413-07 BAKR6413-09 6413-11 6413-09 BAKR6413-07 6413-05
89427-918EA 154.6 USD InStock
CAJT6413-9 CAJT6413-7 89427-918 89427-922 89427-924 89427-920
PTFE [ST] Joint Inserts for Flat Glass Heads, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Solid, virgin PTFE, standard taper joint inserts are designed for use with flat glass heads.
  • Use with flat glass heads
  • Allows for increased working space above the head
  • Secured to head with reverse threaded locking nut
  • Sealed using FETFE O-Ring
This low profile insert allows for increased working space above the head. The insert is secured to the head with a reverse threaded locking nut and sealed using a FETFE O-Ring. The top of the insert is formed to allow the use of certain joint clips. For use with Flat Glass Heads 89054-638 and 89084-094.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty