Would you like to log in again?
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7813-07 7813-11 7813-05 7813-15
89427-980EA 261.13 USD InStock
89427-980 89427-982 89427-978 89427-976
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate storage tube is supplied with an 0-4mm Hi-Vac valve top with a 9mm side port designed to accept ⅜" Cajon fittings
The valved top enables precise flow control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty