Would you like to log in again?
Vacuum Storage Tube with Hi-Vac Top Valve and #15 O-Ring Joint Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7815-05 7815-01 7815-07 7815-03
89427-990EA 298.38 USD InStock
89427-990 89427-988 89427-984 89427-986
Vacuum Storage Tube with Hi-Vac Top Valve and #15 O-Ring Joint Sidearm, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate glass storage tube is supplied with an 0-4mm Hi-Vac valve top with a 15mm O-Ring joint sidearm
The valved top enables precise flow control and the O-Ring joint facilitates connections to Schlenk lines. Supplied with FETFE O-Ring (89061-002).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty