Would you like to log in again?
Vacuum Filter Tube, with Inner Standard Taper Joints on Top and Bottom, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7827-05 7827-03 7827-07 7827-01
89428-002EA 250.97 USD InStock
89428-002 89428-000 89428-006 89428-004
Vacuum Filter Tube, with Inner Standard Taper Joints on Top and Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate glass Schlenk filter tube features an integral glass filter disc at the tube's mid-line, and inner joints at top and bottom.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty