Would you like to log in again?
Vacuum Adapter with Hi-Vac Valved #15 O-Ring Joint Side Port and Ace-Thred Bottom Outlet, Ace Glass Incorporated
Supplier: Ace Glass
15428-28 15428-35 15428-37 15428-33
89428-032EA 214.61 USD InStock
89428-032 89082-828 89428-030 89082-830
Vacuum Adapter with Hi-Vac Valved #15 O-Ring Joint Side Port and Ace-Thred Bottom Outlet, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Vacuum adapters are designed for use with a vacuum cart, vacuum manifold, or Schlenk lines.
  • #15 O-Ring joint side port
  • Hi-Vac valve for precise flow control
  • Ace-Thred bottom outlet
Adapters feature a #15 O-Ring joint side port, with a Hi-Vac valve for precise flow control and an Ace-Thred bottom outlet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty