Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Single Neck Vacuum Flasks with Side Arm, Ace Glass Incorporated
Supplier: Ace Glass
7003-213 7003-215 7003-15 7003-219 7003-29 7003-27 7003-25 7003-11 7003-217 7003-13 7003-211
89428-054EA 261.69 USD InStock
89428-054 89428-042 89428-052 89428-040 89428-036 89428-046 89428-034 89428-044 89428-038 89428-048 89428-050
Single Neck Vacuum Flasks with Side Arm, Ace Glass Incorporated
Flasks Round Bottom Flasks
Round bottom flask with a single neck with standard taper joint and an 8 mm side outlet with either a 2 mm glass stopcock or a 0-3 inlet valve
Ideal for Schlenk lines and either NO-AIR or vacuum applications. Fabricated from borosilicate glass. Includes one 89064-960 septa for side arm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty