Would you like to log in again?
Distillation Packing, Ace Glass Incorporated
Supplier: Ace Glass
6624-09 6624-04 6624-23 6624-28
89428-186EA 1011.76 USD InStock
89428-186 89428-184 89428-182 89428-180
Distillation Packing, Ace Glass Incorporated
Distillation Column Packing
An efficient protruded metal dump-packing for distillation columns of 1" to 12" diameter
  • Self-wetting reduces equilibrium time
  • The dump feature eliminates interlocking, and makes filling and emptying as easy as pouring water
  • Suitable for both atmospheric and reduced pressures down to 10mm Hg condenser pressure
  • Packed heights up to 20 feet are possible without distorting packing
  • Metal packing offered in 316 Stainless Steel and Monel 400
Minimum column diameter for 0.16" packing is 0.75".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty