Would you like to log in again?
Multi Vortex-Genie® Large Capacity Mixer Shaker, Scientific Industries
Supplier: Scientific Industries
Genie
SI-M236 SI-M010 SI-M020
97015-592EA 2754.83 USD InStock
97015-592 89428-532 89428-534
Multi Vortex-Genie® Large Capacity Mixer Shaker, Scientific Industries
Shakers and Mixers Vortex Mixers
A large capacity multi vessel vortex mixer and shaker designed to hold and provide true vortexing in almost any container up to 250ml with the same reliability found in the Vortex-Genie®
Clamping system allows the user to quickly change from one type of vessel to another. Holds multiple test tubes, vials, cylinders of various sizes, or up to six 250ml vessels including Erlenmeyer flasks. Digital control, display of speed and time, Gentle ramp up, may be used in cold rooms and incubators.
Certifications: CE marked.
Ordering information: Two 12-Pack Multi-Tube racks are included. No additional accessories necessary. Two 230V models available - one is supplied without a plug and one is supplied with European plug.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty