Would you like to log in again?
PROSAT® Presaturated Nonwoven Wipers, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROSAT®
PSPP0043 PSPP0043 PSPP0035
89074-010CS 953.39 USD InStock
CA89074-010 89074-010 89428-544
PROSAT® Presaturated Nonwoven Wipers, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Nonwoven wipers designed for cleaning laminar flow workbenches, cleanroom surfaces, and equipment.
Ideal for wiping articles prior to pass through. Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments. Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipers for improved durability, or economical hydroentangled polyester/cellulose wipers.
Ordering information: Packaged in resealable pouches to preserve cleanliness and solvent saturation levels. Pouches are individually lot-numbered for traceability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty