Would you like to log in again?
Kimtech™ A5 Sterile Cleanroom Sleeve Protector, Kimberly-Clark Professional
Supplier: Kimberly-Clark
Kimtech™
36077 36077
89428-610CS 544.78 USD InStock
89428-610 CA89428-610
Kimtech™ A5 Sterile Cleanroom Sleeve Protector, Kimberly-Clark Professional
Disposable Garments
Trust Kimtech™ A5 sterile cleanroom sleeve protectors for a high-value combination of quality and comfort, as well as reliable sterile cleanroom protection.
  • Cuff fold with integrated thumb loop
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms
  • Each individually bagged pair of sleeves has a visual gamma sterilization indicator
  • Not made with natural rubber latex, BHT preservative, silicone
  • Manufactured in an ISO-certified facility, clean-processed, and individually packaged under cleanroom conditions
Kimtech™ A5 sterile cleanroom sleeve protectors have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. They have an innovative cuff fold, which presents the inside of the sleeves as the package is opened, reducing the risk of contaminating the outside of the sleeve. An integrated thumb loop keeps the sleeve from rolling back and exposing the arm or wrist. These protective sleeves are designed for single-use (disposable PPE).
Certifications: CE certified.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty