Would you like to log in again?
VWR® Glass Microanalysis Filter Holders
Supplier: VWR
VWR®
259.202.01 259.202.03
89428-936CS 2379.85 USD InStock
89428-936 89428-934
VWR® Glass Microanalysis Filter Holders
Filter Holders
Used to filter under vacuum/small volumes for particulate or biological contamination analysis and for general filter application.
  • Available in fritted glass or stainless steel
  • Thick pre filters or laminated filters will not seal in this holder
Use durapore (PVDF) membrane or unlaminated PTFE for solvent applications with this apparatus.
Delivery information: The unit is supplied with a coarse porosity fritted or stainless-steel support base, 15 ml graduated funnel, anodized aluminum clamp, and a no.5 silicone stopper.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty