Would you like to log in again?
VWR® Bar-Coded B.O.D. Bottles, 300 ml
Supplier: Avantor
284.455.02Q 284.455.02I 284.457.02 284.455.02P 284.456.02B 284.455.02 284.456.02J 284.456.02H 284.456.02L 284.455.02E 284.455.02K 284.456.02P 284.456.02C 284.456.02K 284.455.02R 284.455.02G 284.456.02Q 284.456.02G 284.456.02N 284.456.02I 284.456.02M 284.456.02O 284.456.02F 284.455.02B 284.455.02H 284.455.02O 284.455.02N
89429-214CS 444.16 USD InStock
89429-214 89429-278 89429-238 89429-284 89429-262 89429-240 89429-282 89429-260 89429-244 89429-266 89429-242 89429-264 89429-220 89429-280 89429-226 89429-268 89429-246 89429-224 89429-228 89429-272 89429-270 89429-210 89429-232 89429-254 89429-276 89429-252 89429-274
VWR® Bar-Coded B.O.D. Bottles, 300 ml
Bottles BOD Bottles
Made from borosilicate glass, these bottles are suitable for determining oxygen demand of sewage water and industrial wastes.
The bottles have a permanently screened label, bar code and are available with interchangeable glass stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty