Would you like to log in again?
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: Eppendorf
Eppendorf® DNA LoBind®
022431048 0030108523 0030108310 022431005 0030122275 0030108523 0030122232 0030122208 0030122348 0030122275 022431021 022431005 0030108310 022431048 022431021
76683-142PK 103.75 USD InStock
CA76683-142 76683-142 470202-584 75871-640 80077-230 CA76683-148 CA89429-316 76683-148 75871-646 89429-316 CA80077-230 CA80077-236 76313-964 80077-236 CA80077-234
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
Unleash the power of precision in DNA research with Eppendorf® DNA LoBind® tubes. Engineered with proprietary LoBind® technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments
  • Precision lid sealing minimizes evaporation rates in the tube format
  • Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000×g (25,000×g for 2.0ml tubes)
  • Snap, screw, and SnapTec® cap options available
Experience unrivaled purity in research with Eppendorf® DNA LoBind® Tubes. Tailored to meet the standards of advanced molecular biology applications, the unparalleled purity grade ensures the integrity of samples. With the innovative LoBind® technology, rest easy knowing that DNA binding is eliminated, safeguarding valuable samples throughout processing. Choose from a variety of cap options (snap, screw, and SnapTec®) to align with experimental requirements, providing the flexibility and convenience needed for any type of work. Whether engaged in PCR, qPCR, or DNA sequencing, the thoughtfully designed cap options enhance the overall user experience, putting control and precision at your fingertips.
Applications include preparation or storage of DNA and RNA samples, forensic trace analysis, preparing dilution series in quantitative qPCR, sample Preparation in next-generation sequencing, and creation of genomic or oligonucleotide libraries.
Manufactured without the use of suface coatings, slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Certifications: ACT label (5 and 25 ml), Lot-specific certificates can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty