Would you like to log in again?
Sabouraud Dextrose (SabDex) Agar Blue, Hardy Diagnostics
Supplier: Hardy Diagnostics
W82 W82
89429-348PK 23.66 USD InStock
CA89429-348 89429-348
Sabouraud Dextrose (SabDex) Agar Blue, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose (SabDex) Agar, Blue, not for human diagnostic use, 26ml fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
This product is for laboratory use only; not for human diagnostic use. See VWR catalog number 89407-894 for Sabdex Agar that is intended for human diagnostic use.
This product contains an inert blue dye to facilitate differentiation from other media that would otherwise have an similar color (such as Tryptic Soy Agar).
(Sabourauds Dextrose Agar) For the cultivation of fungi.
This product contains an inert blue dye to facilitate differentiation from other media that would otherwise have an similar color (such as Tryptic Soy Agar).
Certifications: USP
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty