Would you like to log in again?
Middlebrook 7H11 Selective, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W40 W40
89429-350PK 72.31 USD InStock
89429-350 CA89429-350
Middlebrook 7H11 Selective, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Middlebrook 7H11 Selective, for mycobacteria, deep fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
(Mitchison formula with CAPT antibiotics) For selective isolation of Mycobacterium spp.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty