Would you like to log in again?
Wilkins-Chalgren Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
G89 G89
89429-412PK 93.55 USD InStock
89429-412 CA89429-412
Wilkins-Chalgren Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Wilkins-Chalgren Agar, 15x100mm plate, order by the package of 10, by Hardy Diagnostics.
Hardy Diagnostics Wilkins-Chalgren Agar is recommended for the cultivation and isolation of anaerobic microorganisms and for the preparation of agar dilution tests.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty