Would you like to log in again?
Hygiena SuperSnap™, High-Sensitivity ATP surface test, Hardy Diagnostics
Supplier: Hardy Diagnostics
SuperSnap™
SUS3000 SUS3000
89492-624EA 461.61 USD InStock
CA89492-624 89492-624
Hygiena SuperSnap™, High-Sensitivity ATP surface test, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
SuperSnap™ is the most sensitive ATP surface test in Hygiena's ATP hygiene monitoring portfolio of products. SuperSnap™ is an all-in-one test which detects extremely low levels of ATP (Adenosine Triphosphate).
  • Sensitivity - Up to 100x better than other systems
  • Allergen prevention
  • Process monitoring for perishable/short-life foods and beverages
  • Industrial applications with strict cleaning protocols
  • Improved detection at very low ATP values (e.g. dairies and beverage processors)
  • Improved Buffer Capacity
  • Extremely resistant to harsh or concentrated chemical sanitizers
  • Works with samples that have extremely high and low pH - 5 times more resistant to acid, 10 times more resistant to alkali
  • Decreases wait time to test by allowing sample collection on wet sanitized surfaces
Used predominantly in Food & Beverage processing facilities as an allergen cross-contamination prevention tool or when dealing with difficult samples, SuperSnap™ can identify food residues at levels similar to protein levels detected by specific allergen tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty