Would you like to log in again?
Cryostore™ Storage Boxes for Cryogenic Vials, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CryoStore™
T314-225G T314-581R T314-225R T314-281G T314-281Y T314-225B T314-481B T314-225B T314-281R T314-2100R T314-542Y T314-542Y T314-281B T314-2100B T314-542R T314-225Y T314-481B T314-225R T314-481R T314-225B T314-581Y T314-581B T314-481G T314-542R T314-481Y T314-2100R T314-581Y T314-2100B T314-225G T314-2100G T314-481R T314-542B T314-581R T314-481Y T314-2100Y T314-281B T314-281R T314-481G T314-581G T314-2100Y T314-542B T314-581G T314-542G T314-225Y T314-581B T314-542G T314-2100G
89492-694CS 215.97 USD InStock
CA48648-113 CA48648-112 CA48648-111 CA48648-110 CA11007-768 89492-694 CA11006-980 89492-672 89492-674 89492-696 490014-848 89492-698 89492-676 89176-988 89492-678 CA11006-978 CA11006-974 89492-690 CA11006-976 89492-670 89492-692 CA48648-108 CA48648-106 89492-702 89176-992 89176-994 89492-704 CA11028-530 89176-990 89492-684 CA11006-970 89492-686 CA11006-972 89492-688 89492-666 89492-668 89492-700 CA11006-966 CA11008-454 CA11006-968 CA11008-456 89492-680 CA11008-458 89492-682 CA11028-524 CA11028-528 CA11028-526
Cryostore™ Storage Boxes for Cryogenic Vials, Simport Scientific
Boxes Cryoboxes
Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196 °C and +121 °C and are autoclavable at 120 °C, 15 psig (1 bar) for 20 minutes
Colored plastic grids separate the cover from the base on the 25, 42 and 81-place boxes. Those made to accept 100 tubes have a colored base instead of a grid.
A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Printed with a series of squares (numbered from 1 to 25, 1 to 42, 1 to 81, or 1 to 100), surface accepts writing with markers, facilitating inventory control.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty