Would you like to log in again?
Cryovial® Workstation Rack for Cryogenic Vials, Simport Scientific
Supplier: Simport
T315-10 T315-2 T315-2 T315-3 T315-3 T315-10
89492-706CS 55.45 USD InStock
CA48648-123 CA48648-121 89492-706 CA48648-120 89492-708 89492-710
Cryovial® Workstation Rack for Cryogenic Vials, Simport Scientific
Racks Cryogenic Vial Racks
This handy autoclavable rack can hold up to 50 to 52 cryogenic vials.
Made of polypropylene.
Thanks to an innovative universal locking system, the vials will securely lock in each well and will not turn. Each position is identified with an alphanumeric index. Strong handles make it easy and safe to carry. It is supported by anti-skid rubber feet. The rack is compact and stackable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty