Would you like to log in again?
Porcelain Crucibles, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1883-06 CG-1882-04 CG-1882-02 CG-1883-05 CG-1882-06 CG-1883-07 CG-1882-01 CG-1883-02 CG-1883-01 CG-1882-05 CG-1883-04 CG-1883-03 CG-1882-03
89492-772EA 10.92 USD InStock
89492-772 89492-794 89492-782 89492-774 89492-784 89492-776 89492-786 89492-778 89492-788 89492-790 89492-792 89492-770 89492-780
Porcelain Crucibles, Chemglass
Crucibles
Porcelain crucible complete with lid. Glazed both inside and outside, with the exception of the outside lower bottom and rim.
Porcelain crucibles are widely used in the chemistry laboratory
The average coefficient of expansion from 20 to 200°C is 3.56 x 10-5 gradually increasing to 4.69 x 10-5 at 1000°C. To prevent thermal stress cracks on porcelain ware, do not exceed a heating/cooling rate of 200°C per hour.
They offer excellent resistance to all reagents except hydrofluoric acid, and are highly resistant to thermal shock while having a very low coefficient of expansion.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty